Anti-Procathepsin K Rabbit Polyclonal Antibody

Supplier: BioVision
BIOV6667-100EA 299 EUR
BIOV6667-100
Anti-Procathepsin K Rabbit Polyclonal Antibody
Antibodies
Cathepsin K is a member of the papain cysteine proteinase family and has been identified as the predominant proteinase responsible for the resorption of the bone matrix. The enzyme cleaves proteins such as collagen type I, collagen type II and osteonectin and therefore plays a role in bone remodeling and resorption in diseases such as osteoporosis, osteolytic bone metastasis and rheumatoid arthritis (Bromme and Okamoto, 1995; Drake, F. et al 1996; Bossard et al, 1996;

Recommended Dilutions: Western Blot: 1-4 µg/ml.

Type: Primary
Antigen: Procathepsin K
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR