VWR® HPLC and GC Certified Vial Kits
Supplier: VWR
VWR®
548-1488EA 0 EUR
548-1488
VWR® HPLC and GC Certified Vial Kits
Vials
The use of HPLC and GC certified vial kits are an important way of reducing sources of error and ensuring that processes are reproducible.
  • Each batch of HPLC and GC certified kits is tested on 15 critical parameters. In a realistic method, an HPLC/UV and GC-MS test of vials/closure combination of blanks and contaminations will be carried out
  • The batch-specific test certificate, with the HPLC and GC chromatograms, can be handed out upon request
Packaging: HPLC and GC certified kits are supplied completely shrink-wrapped to ensure origin, purity and transport safety.