LC-MS and GC-MS certified vial kits
548-1990EA 821 EUR
VWRI548-1990 VWRI548-1991
LC-MS and GC-MS certified vial kits
Each vial/closure combination lot has been tested by LC-MS and GC-MS on traces of blank values and contaminations.
  • Additionally the glass surface of these specific SureStop™ vials provides very low adsorption tendencies for all types of polar compounds
  • Closures contain a very soft ultra-low bleed (Ultra High Performance) silicone septum with PTFE layer, optimised for ultra-trace analysis
  • The batch-specific test certificate with the MS chromatograms can be handed out on request
Packaging: The LC-MS and GC-MS certified kits are delivered completely shrink-wrapped in order to assure origin, purity and transport safety.