Anti-ZBED1 Rabbit Polyclonal Antibody

Supplier: Bioss
BOSSBS-13030REA 691 EUR
BOSSBS-13030R
Anti-ZBED1 Rabbit Polyclonal Antibody
Antibodies
ZBED1 is a 694 amino acid protein that localizes specifically to granular structures within the nucleus. Expressed ubiquitously at low levels and present at higher levels in heart, placenta, spleen and skeletal muscle, ZBED1 is thought to function as a transcription factor that regulates a number of ribosomal protein (RP) encoding genes, thereby playing a role in the cell cycle and in cell proliferation events. ZBED1 contains one BED-type zinc finger and binds specifically to 5'-TGTCG[CT]GA[CT]A-3' DNA regions found in RP promotors. Additionally, ZBED1 binds strongly to the promotor region of Histone H1 (a protein required for the condensation of nucleosomes into higher order structures), subsequently activating H1 transcription.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: ZBED1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR