Anti-GPR12 Rabbit Polyclonal Antibody

Supplier: Bioss
BOSSBS-13514REA 691 EUR
BOSSBS-13514R
Anti-GPR12 Rabbit Polyclonal Antibody
Antibodies
G protein-coupled receptors (GPRs) are a protein family of transmembrane receptors that transmit an extracellular signal (ligand binding) into an intracellular signal (G protein activation). GPR signaling is an evolutionarily ancient mechanism used by all eukaryotes to sense environmental stimuli and mediate cell-cell communication. All of the receptors have seven membrane-spanning domains and the extracellular parts of the receptor can be glycosylated. These extracellular loops also contain two highly conserved cysteine residues which create disulfide bonds to stabilize the receptor structure. GPR12 is a 334 amino acid peptide that is expressed primarily in brain, particularly in regions where neuronal differentiation takes place. GPR12 is coupled to an inhibitory G protein. It positively influences differentiation and maturation of post-mitotic neurons, and it may promote the growth of neuronal precursor cells.

WB(1:100-1000), FCM(1:20-100), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: GPR12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR