Anti-pan methyl Lysine Rabbit Polyclonal Antibody

Supplier: Bioss
BOSSBS-6350REA 691 EUR
BOSSBS-6350R
Anti-pan methyl Lysine Rabbit Polyclonal Antibody
Antibodies
Lysine methylation occurs in three distinct states, having either one (me1), two (me2) or three (me3) methyl groups attached to the amine group of the lysine side chain. In eukaryotes, histone H3 trimethylated at lysine 4 (H3K4me3) is associated with active chromatin and gene expression.

IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: pan methyl Lysine
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR