Anti-ZDHHC12 Rabbit Polyclonal Antibody

Supplier: Bioss
BOSSBS-6958REA 691 EUR
BOSSBS-6958R
Anti-ZDHHC12 Rabbit Polyclonal Antibody
Antibodies
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. ZDHHC12 (zinc finger, DHHC domain containing 12), also known as ZNF400, is a 267 amino acid multi-pass membrane protein that contains one DHHC-type zinc finger and is thought to function as a palmitoyltransferase, catalyzing the transformation of palmitoyl-CoA and a cysteine-conjugated protein to a S-palmitoyl protein and free CoA. Three isoforms exist due to alternative splicing events. The gene encoding ZDHHC12 maps to human chromosome 9, which consists of about 145 million bases and 4% of the human genome and encodes nearly 900 genes. Chromosome 9 is considered to play a role in gender determination, deletion of the distal portion of 9p can lead to development of male to female sex reversal, the phenotype of a female with a male X,Y genotype.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: ZDHHC12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR