Anti-PHF8 Rabbit Polyclonal Antibody

Supplier: Bioss
BOSSBS-8176REA 691 EUR
BOSSBS-8176R
Anti-PHF8 Rabbit Polyclonal Antibody
Antibodies
The protein encoded by this gene is a histone lysine demethylase that preferentially acts on histones in the monomethyl or dimethyl states. The encoded protein requires Fe(2+) ion, 2-oxoglutarate, and oxygen for its catalytic activity. Defects in this gene are a cause of mental retardation syndromic X-linked Siderius type (MRXSSD). Four transcript variants encoding different isoforms have been found for this gene.

IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: PHF8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR