(-)-4,5-O-Cyclohexylidenequinic acid lactone for synthesis
Supplier: Merck
M-Clarity™ quality level = MQ200
8.18241.0005EA
150
EUR
8.18241.9999
8.18241.0005
(-)-4,5-O-Cyclohexylidenequinic acid lactone for synthesis
(-)-4,5-O-Cyclohexylidenequinic acid lactone
Formula:
C₁₃H₁₈O₅ |
CAS Number:
35949-53-2 |
Specification Test Results
Solubility | (20 °C) insoluble |
Melting Point | 138 - 141 °C |
Molar Mass | 254.28 g/mol |
Bulk Density | 210 kg/m3 |
Hill Formula | C13H18O5 |
Storage | +15...+25 °C |
Assay (DSC) | ≥99,0% |
Specific rotation | −34,0 - −33,0 ° |
Identity (IR) | Passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...