Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-WWTR1 Rabbit Polyclonal Antibody

Supplier: Abgent
Type: Primary
Antigen: WWTR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Pk VWR Catalog Number Quantity
400 µl ABGEAP16419B
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol WWTR1
Antigen name WW domain containing transcription regulator 1
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Western blot Yes
ELISA Yes
Antigen synonyms TAZ
Amino acid number 370 to 398
Gene ID 25937
Epitope C-terminal
Concentration 0.55
Immunogen This WWTR1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 370-398 amino acids from the C-terminal region of human WWTR1.
Molecular weight 44.101 kDa
Purification Protein A & Peptide affinity purification
Sequence DVESALNKSEPFLTC
Storage buffer pH 7,4
Storage temperature Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at – 20 °C in small aliquots to prevent freeze-thaw cycles
Shipping temperature 4 °C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty