Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ZNF350 Rabbit Polyclonal Antibody

Supplier: Abgent
Type: Primary
Antigen: ZNF350
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human




 

ORDER

Expand All / Collapse All
Pk VWR Catalog Number Quantity
400 µl ABGEAP16724C
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol ZNF350
Antigen name zinc finger protein 350
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Western blot Yes
ELISA Yes
Antigen synonyms ZBRK1, ZFQR
Amino acid number 176 to 204
Gene ID 59348
Epitope internal
Concentration 0.95
Immunogen This ZNF350 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 176-204 amino acids from the Central region of human ZNF350.
Molecular weight 60.011 kDa
Purification Protein A & Peptide affinity purification
Sequence FISPKHQKTRKLEKC
Storage buffer pH 7,4
Storage temperature Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at – 20 °C in small aliquots to prevent freeze-thaw cycles
Shipping temperature 4 °C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty