Anti-PROK1 Rabbit Polyclonal Antibody

Supplier: Boster
BSBTPB9881EA 579 EUR
BSBTPB9881
Anti-PROK1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PROK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR