Anti-SMYD3 Rabbit Polyclonal Antibody

Supplier: Boster
BSBTPB9893EA 550 EUR
BSBTPB9893
Anti-SMYD3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SMYD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR