Ammonium hexachloroiridate(III) hydrate
Supplier: Thermo Scientific
|
Warning
|
12632.06EA
1900
EUR
12632.06
Ammonium hexachloroiridate(III) hydrate
Ammonium hexachloroiridate(III) hydrate
Formula:
(NH₄)₃IrCl₆·xH₂O Storage Temperature: Ambient |
MDL Number:
MFCD00003393 CAS Number: 15752-05-3 EINECS: 239-842-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...