Potassium hydrogen diiodate 0.025 N volumetric solution

Supplier: Thermo Scientific

Synonyms: Potassium bi-iodate

35600.K2EA 0 EUR
35600.K2
Potassium hydrogen diiodate 0.025 N volumetric solution
Potassium hydrogen diiodate
Formula: HI₂KO₆
MW: 389,91
Storage Temperature: Ambient
MDL Number: MFCD00011400
CAS Number: 13455-24-8
EINECS: 236-650-9
UN: 1479
ADR: 5.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR