Ruthenium 4% on alumina, rings
Supplier: Thermo Scientific
44593.36EA
2410
EUR
44593.36
Ruthenium 4% on alumina, rings
Ruthenium
Formula:
Ru MW: 101,07 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011207 CAS Number: 7440-18-8 EINECS: 231-127-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...