2,4,6-Trimethoxyboroxin

Supplier: Thermo Scientific
Danger

Synonyms: TrimethoxycyclotriboroxaneTrimethoxyboroxine

89153.14EA 27 EUR
89153.14
2,4,6-Trimethoxyboroxin
2,4,6-Trimethoxyboroxin
Formula: C₃H₉B₃O₆
MW: 173,53 g/mol
Boiling Pt: 130 °C
Melting Pt: 10…11 °C
Density: 1,216
Flash Pt: 14 °C (57 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005946
CAS Number: 102-24-9
EINECS: 203-016-8
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR