3,5-Diiodo-L-tyrosine dihydrate, (max. 10% H₂O) 98% (dry weight)

Supplier: Thermo Scientific
Warning

A10702.22EA 223 EUR
A10702.22 A10702.06 A10702.14
3,5-Diiodo-L-tyrosine dihydrate, (max. 10% H₂O) 98% (dry weight)
3,5-Diiodo-L-tyrosine dihydrate
Formula: C₉H₉I₂NO₃
Melting Pt: 185 °C (dec.)
MDL Number: MFCD00016542
CAS Number: 18835-59-1
EINECS: 206-092-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR