1-(2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)ethanone 98%

Supplier: Thermo Scientific

A15506.14EA 155 EUR
A15506.14 A15506.06 A15506.22
1-(2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)ethanone 98%
1-(2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)ethanone
Formula: C₁₀H₁₀O₃
MW: 178,19
Melting Pt: 81…84 °C
Storage Temperature: Ambient
MDL Number: MFCD00006823
CAS Number: 2879-20-1
EINECS: 220-726-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR