Ethyl-3-methyl-4-nitrobenzoate 98%

Supplier: Thermo Scientific

A18625.09EA 24.7 EUR
A18625.09 A18625.30 A18625.18
Ethyl-3-methyl-4-nitrobenzoate 98%
Ethyl-3-methyl-4-nitrobenzoate
Formula: C₁₀H₁₁NO₄
MW: 209,20
Melting Pt: 53…55 °C
Storage Temperature: Ambient
MDL Number: MFCD00115804
CAS Number: 30650-90-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR