2,6,8-Trimethyl-4-nonanol (threo- and erythro- mixture) ≥90%

Supplier: Thermo Scientific

Synonyms: 4-Hydroxy-2,6,8-trimethylnonane

B20830.14EA 229 EUR
B20830.14
2,6,8-Trimethyl-4-nonanol (threo- and erythro- mixture) ≥90%
2,6,8-Trimethyl-4-nonanol (threo- and erythro- mixture)
Formula: C₁₂H₂₆O
Boiling Pt: 123 °C (20 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00026491
CAS Number: 123-17-1
EINECS: 204-606-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR