(9,9-Dimethyl-9H-xanthene-4,5-diyl)bis(diphenylphosphine) 97%

Supplier: Thermo Scientific

Synonyms: XANTPHOS4,5-Bis(diphenylphosphino)-9,9-dimethylxanthene9,9-Dimethyl-4,5-bis(diphenylphosphino)xanthene

H26241.06EA 238 EUR
H26241.06 H26241.03
(9,9-Dimethyl-9H-xanthene-4,5-diyl)bis(diphenylphosphine) 97%
(9,9-Dimethyl-9H-xanthene-4,5-diyl)bis(diphenylphosphine)
Formula: C₃₉H₃₂OP₂
MW: 578,62 g/mol
Melting Pt: 224…228 °C
Storage Temperature: Ambient
MDL Number: MFCD00233866
CAS Number: 161265-03-8
EINECS: 605-249-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR