4-(Trifluoromethyl)cyclohexanecarboxylic acid (cis and trans mixture) ≥97%

Supplier: Thermo Scientific

Synonyms: 4-(Trifluoromethyl)cyclohexane-1-carboxylic acid

H27283.03EA 0 EUR
H27283.03
4-(Trifluoromethyl)cyclohexanecarboxylic acid (cis and trans mixture) ≥97%
4-(Trifluoromethyl)cyclohexanecarboxylic acid (cis and trans mixture)
Formula: C₈H₁₁F₃O₂
MW: 196,17 g/mol
Melting Pt: 88…94 °C
MDL Number: MFCD00220049
CAS Number: 95233-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR