4-Pyrimidinemethanol 98%

Supplier: Thermo Scientific
Warning

Synonyms: 4-(Hydroxymethyl)pyrimidine

H31754.MDEA 207 EUR
H31754.MD
4-Pyrimidinemethanol 98%
4-Pyrimidinemethanol
Formula: C₅H₆N₂O
MW: 110,12 g/mol
Melting Pt: 75…78 °C
Storage Temperature: Ambient
MDL Number: MFCD03789625
CAS Number: 33581-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR