2-(N-Benzyl-N-methylamino)-5-bromopyridine 95%

Supplier: Thermo Scientific
Warning

H32929.03EA 152 EUR
H32929.03
2-(N-Benzyl-N-methylamino)-5-bromopyridine 95%
2-(N-Benzyl-N-methylamino)-5-bromopyridine
Formula: C₁₃H₁₃BrN₂
Storage Temperature: Ambient
MDL Number: MFCD12913988
CAS Number: 1187386-36-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR