3,5,6-Trifluoro-2-hydroxypyridine 98%

Supplier: Thermo Scientific
Warning

Synonyms: 3,5,6-Trifluoro-2-pyridinol3,5,6-Trifluoro-2-pyridone

H33085.03EA 102 EUR
H33085.03
3,5,6-Trifluoro-2-hydroxypyridine 98%
3,5,6-Trifluoro-2-hydroxypyridine
Formula: C₅H₂F₃NO
MW: 149,07 g/mol
Melting Pt: 99…103 °C
MDL Number: MFCD04114299
CAS Number: 75777-49-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR