(R)-(+)-5,5',6,6',7,7',8,8'-Octahydro-1,1'-bi-2-naphthol ≥98%, ee 99%

Supplier: Thermo Scientific
Warning

Synonyms: (R)-(+)-2,2'-Dihydroxy-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthyl

H36142.03EA 92.6 EUR
H36142.03 H36142.06
(R)-(+)-5,5',6,6',7,7',8,8'-Octahydro-1,1'-bi-2-naphthol ≥98%, ee 99%
(R)-(+)-5,5',6,6',7,7',8,8'-Octahydro-1,1'-bi-2-naphthol
Formula: C₂₀H₂₂O₂
MW: 294,39 g/mol
Melting Pt: 160…166 °C
Storage Temperature: Ambient
MDL Number: MFCD02093485
CAS Number: 65355-14-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR