(S)-(-)-5,5',6,6',7,7',8,8'-Octahydro-1,1'-bi-2-naphthol ≥98%, ee 99%

Warning

Synonyms: (S)-5,5,6,6,7,7,8,8-Octahydro-1,1-bi-2-naphthol(S)-(-)-2,2'-Dihydroxy-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthyl

H37716.03EA 94.9 EUR
H37716.03 H37716.06
(S)-(-)-5,5',6,6',7,7',8,8'-Octahydro-1,1'-bi-2-naphthol ≥98%, ee 99%
(S)-(-)-5,5',6,6',7,7',8,8'-Octahydro-1,1'-bi-2-naphthol
Formula: C₂₀H₂₂O₂
MW: 294,39 g/mol
Melting Pt: 160…166 °C
Storage Temperature: Ambient
MDL Number: MFCD02093485
CAS Number: 65355-00-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR