(3-Carbamoylphenyl)boronic acid 98%
|
Danger
|
Synonyms:
3-Carbamoylphenylboronic acid
, 3-Carbamoylbenzeneboronic acid
, 3-carbamoylbenzeneboronic acid
, (3-carbamoylphenyl)boronic acid
, 3-Aminocarbonylphenylboronic acid
H52869.06EA
192
EUR
H52869.06
(3-Carbamoylphenyl)boronic acid 98%
(3-Carbamoylphenyl)boronic acid
Formula:
C₇H₈BNO₃ MW: 164,955 g/mol Melting Pt: 232…238 °C Storage Temperature: Ambient |
MDL Number:
MFCD03411948 CAS Number: 351422-73-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...