Methyl-3,5-diamino-6-chloropyrazine-2-carboxylate 98%

Supplier: Thermo Scientific
Warning

Synonyms: Methyl 6-chloro-3,5-diaminopyrazine-2-carboxylate

H55657.03EA 65.6 EUR
H55657.03 H55657.06
Methyl-3,5-diamino-6-chloropyrazine-2-carboxylate 98%
Methyl-3,5-diamino-6-chloropyrazine-2-carboxylate
Formula: C₆H₇ClN₄O₂
MW: 202,60
Melting Pt: 211…215 °C
Storage Temperature: Ambient
MDL Number: MFCD01928388
CAS Number: 1458-01-1
EINECS: 215-946-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR