Piperidin-3-amine dihydrochloride 95%

Supplier: Thermo Scientific
Warning

Synonyms: piperidin-3-amine dihydrochloride3-Aminopiperidine dihydrochloride3-Amino-Piperidine Di-Hydrochloride

H57306.03EA 570 EUR
H57306.03 H57306.MD
Piperidin-3-amine dihydrochloride 95%
Piperidin-3-amine dihydrochloride
Formula: C₅H₁₄Cl₂N₂
MW: 173,081 g/mol
Melting Pt: 230 °C
Storage Temperature: Ambient
MDL Number: MFCD00012773
CAS Number: 138060-07-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR