p-Toluenesulphonylsemicarbazide 95%
Supplier: Thermo Scientific
|
Warning
|
H61949.22EA
344
EUR
H61949.22
H61949.14
p-Toluenesulphonylsemicarbazide 95%
p-Toluenesulphonylsemicarbazide
Formula:
C8H11N3O3S MW: 229,26 g/mol |
MDL Number:
MFCD00072243 CAS Number: 10396-10-8 EINECS: 233-857-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...