4-Chloro-2-fluoro-5-nitrobenzoic acid 98%
|
Warning
|
H63760.06EA
189
EUR
H63760.06
H63760.MD
4-Chloro-2-fluoro-5-nitrobenzoic acid 98%
4-Chloro-2-fluoro-5-nitrobenzoic acid
Formula:
C₇H₃ClFNO MW: 219,55 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04115725 CAS Number: 35112-05-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...