(R)-Morpholin-2-ylmethanol hydrochloride 95%

Supplier: Thermo Scientific
Warning

Synonyms: (R)-2-Morpholinemethanol hydrochloride

H64003.03EA 607 EUR
H64003.03
(R)-Morpholin-2-ylmethanol hydrochloride 95%
(R)-Morpholin-2-ylmethanol hydrochloride
Formula: C₅H₁₁NO₂·HCl
MW: 153,61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11052541
CAS Number: 1436436-17-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR