2-Chloro-5-(trifluoromethoxy)phenylboronic acid 98%
|
Warning
|
Synonyms:
(2-Chloro-5-(trifluoromethoxy)phenyl)boronic acid
, 2-Chloro-5-(trifluoromethoxy)benzeneboronic acid
H64341.06EA
126
EUR
H64341.06
H64341.03
2-Chloro-5-(trifluoromethoxy)phenylboronic acid 98%
2-Chloro-5-(trifluoromethoxy)phenylboronic acid
Formula:
C₇H₅BClF₃O₃ MW: 240,37 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06656267 CAS Number: 1022922-16-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...