Methyl-3-hydroxy-4-iodobenzoate 97%

Supplier: Thermo Scientific
Warning

H64354.06EA 343 EUR
H64354.06 H64354.MD H64354.03
Methyl-3-hydroxy-4-iodobenzoate 97%
Methyl-3-hydroxy-4-iodobenzoate
Formula: C₈H₇IO₃
MW: 278,05
Storage Temperature: Ambient
MDL Number: MFCD06203960
CAS Number: 157942-12-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR