5-Fluoro-3-nitropyridin-2(1H)-one 98%

Supplier: Thermo Scientific
Warning

Synonyms: 5-Fluoro-2-hydroxy-3-nitropyridine

H64441.03EA 65.3 EUR
H64441.03 H64441.06 H64441.MD
5-Fluoro-3-nitropyridin-2(1H)-one 98%
5-Fluoro-3-nitropyridin-2(1H)-one
Formula: C5H3FN2O3
MW: 158,09
Storage Temperature: Ambient
MDL Number: MFCD05662412
CAS Number: 136888-20-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR