2-Bromodibenzofuran 98%

Supplier: Thermo Scientific
Warning

H64596.03EA 0 EUR
H64596.03 H64596.MD
2-Bromodibenzofuran 98%
2-Bromodibenzofuran
Formula: C₁₂H₇BrO
MW: 247,09 g/mol
Melting Pt: 108…111 °C
Storage Temperature: Ambient
MDL Number: MFCD00092338
CAS Number: 86-76-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR