3-Fluoro-2-methox-6-methylypyridine 97%

Supplier: Thermo Scientific
Warning

Synonyms: 3-Fluoro-2-methoxy-6-picoline

H64683.03EA 26.9 EUR
H64683.03 H64683.06
3-Fluoro-2-methox-6-methylypyridine 97%
3-Fluoro-2-methox-6-methylypyridine
Formula: C₇H₈FNO
MW: 141,15
Storage Temperature: Ambient
MDL Number: MFCD03095083
CAS Number: 375368-80-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR