3-Chloro-2-fluoro-5-(trifluoromethyl)pyridine 98%

Supplier: Thermo Scientific
Warning

H64723.03EA 24.2 EUR
H64723.03 H64723.14 H64723.06
3-Chloro-2-fluoro-5-(trifluoromethyl)pyridine 98%
3-Chloro-2-fluoro-5-(trifluoromethyl)pyridine
Formula: C₆H₂ClF₄N
MW: 199,53
Boiling Pt: 50…55 °C (11 mmHg)
Melting Pt: 16…20 °C (dec.)
Density: 1,524
Flash Pt: 52 °C (126 °F)
Storage Temperature: Ambient
MDL Number: MFCD00191918
CAS Number: 72537-17-8
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR