2,4-Dibromobenzyl alcohol 98%

Supplier: Thermo Scientific
Warning

H64741.MDEA 29.3 EUR
H64741.MD
2,4-Dibromobenzyl alcohol 98%
2,4-Dibromobenzyl alcohol
Formula: C₇H₆Br₂O
Storage Temperature: Ambient
MDL Number: MFCD09996906
CAS Number: 666747-06-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR