2-Chloro-N-fmoc-L-phenylalanine 97%
Supplier: Thermo Scientific
Synonyms:
Fmoc-Phe(2-Cl)-OH
H66258.06EA
105
EUR
H66258.06
2-Chloro-N-fmoc-L-phenylalanine 97%
2-Chloro-N-fmoc-L-phenylalanine
Formula:
C₂₄H₂₀ClNO₄ MW: 421,877 g/mol |
MDL Number:
MFCD00797578 CAS Number: 198560-41-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...