2-Amino-6-methoxypyridine 97%

Warning

H66436.14EA 313 EUR
H66436.14 H66436.06
2-Amino-6-methoxypyridine 97%
2-Amino-6-methoxypyridine
Formula: C₆H₈N₂O
MW: 124,14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04972542
CAS Number: 17920-35-3
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR