Pig Glutamic oxalacetic transaminase (from Heart)
Supplier: Thermo Fisher Scientific
J60044.4IEA 22.3 EUR
ALFAJ60044.4I
Pig Glutamic oxalacetic transaminase (from Heart)
Applications: Converts L-aspartate and alpha-ketoglutarate to oxaloacetate and L-glutamate.