FSB buffer

Supplier: Thermo Fisher Scientific
J60262.AKEA 158 EUR
J60262.AK
FSB buffer
Buffers Cell Culture Buffers
Storage temperature: 2 to 8 °C
Physical form: Liquid
Appearance: Colourless
Applications: For frozen storage of competent cells
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR