Astemizole ≥99%
Supplier: Thermo Scientific
|
Warning
|
J60339.MFEA
191
EUR
J60339.MF
Astemizole ≥99%
Astemizole
Applications: Selective histamine H1 receptor antagonist
Formula:
C₂₈H₃₁FN₄O MW: 458,57 g/mol Melting Pt: 149…150 °C |
MDL Number:
MFCD00153919 CAS Number: 68844-77-9 EINECS: 272-441-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...