1,2,3,4,5,6-Hexabromocyclohexane
Supplier: Thermo Scientific
Synonyms:
Benzene Hexabromide
J60853.06EA
465
EUR
J60853.06
J60853.03
1,2,3,4,5,6-Hexabromocyclohexane
1,2,3,4,5,6-Hexabromocyclohexane
Applications: Potently and directly inhibits JAK2 tyrosine kinase autophosphorylation
Formula:
C₆H₆Br₆ MW: 557,54 g/mol |
MDL Number:
MFCD00059127 CAS Number: 1837-91-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...