Carbonate-buffered saline (5X), pH 9,0

Supplier: Thermo Fisher Scientific
J61396.AKEA 41.4 EUR
J61396.AK J61396.AP
Carbonate-buffered saline (5X), pH 9,0
Buffers Cell Culture Buffers
CAS No.: 497-19-8
Physical Form: Liquid
MDL No.: MFCD00003494
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR