2-Amino-6-chloropurine riboside 95%

Supplier: Thermo Scientific

Synonyms: 2-Amino-6-chloro-9-(beta-D-ribofuranosyl)purine(2R,3R,4S,5R)-2-(2-amino-6-chloro-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol

J64468.03EA 86.9 EUR
J64468.03
2-Amino-6-chloropurine riboside 95%
2-Amino-6-chloropurine riboside
Formula: C₁₀H₁₂ClN₅O₄
MW: 301,69 g/mol
Melting Pt: 165…167 °C
MDL Number: MFCD00005735
CAS Number: 2004-07-1
EINECS: 217-905-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR