2'-O-Methyluridine 99%

Supplier: Thermo Scientific

J64897.03EA 29.8 EUR
J64897.03
2'-O-Methyluridine 99%
2'-O-Methyluridine
Formula: C₁₀H₁₄N₂O₆
MDL Number: MFCD00056054
CAS Number: 2140-76-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR